4-Fluor-2-nitroanisol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluoro-1-methoxy-2-nitrobenzene

PC3883-25G PC3883-5G PC3883-100G
APOSPC3883-25GEA 29 EUR
APOSPC3883-25G APOSPC3883-5G APOSPC3883-100G
4-Fluor-2-nitroanisol 98%
4-Fluor-2-nitroanisol
Formel: C₇H₆FNO₃
Molecular Weight: 171,13 g/mol
Schmelzpunkt: 62…64 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013375
CAS-Nummer: 445-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR