2-Fluor-4-nitrobenzoesäure 97%

Lieferant: Apollo Scientific
Achtung

PC3912-5G
APOSPC3912-5GEA 21.9 EUR
APOSPC3912-5G
2-Fluor-4-nitrobenzoesäure 97%
2-Fluor-4-nitrobenzoesäure
Formel: C₇H₄FNO₄
Molecular Weight: 185,11 g/mol
Schmelzpunkt: 172…177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00275565
CAS-Nummer: 403-24-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR