2-Fluor-5-nitrobenzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC3931-100G
PC3931-25G
PC3931-5G
APOSPC3931-100GEA
0
EUR
APOSPC3931-100G
APOSPC3931-25G
APOSPC3931-5G
2-Fluor-5-nitrobenzoesäure 98%
2-Fluor-5-nitrobenzoesäure
Formel:
C₇H₄FNO₄ Schmelzpunkt: 139…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134238 CAS-Nummer: 7304-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...