(2-Fluorophenyl)glycine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-2-(2-fluorophenyl)acetic acid
PC3964-5G
PC3964-1G
PC3964-10G
APOSPC3964-5GEA
215
EUR
APOSPC3964-5G
APOSPC3964-1G
APOSPC3964-10G
(2-Fluorophenyl)glycine 97%
(2-Fluorophenyl)glycine
Formel:
C₈H₈FNO₂ Molecular Weight: 169,16 g/mol Schmelzpunkt: 290 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042726 CAS-Nummer: 84145-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...