6-Fluorpicolinsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-6-pyridinecarboxylic acid, 6-Fluoropyridine-2-carboxylic acid
PC3966-1G
PC3966-5G
PC3966-10G
APOSPC3966-1GEA
21.9
EUR
APOSPC3966-1G
APOSPC3966-5G
APOSPC3966-10G
6-Fluorpicolinsäure 98%
6-Fluorpicolinsäure
Formel:
C₆H₄FNO₂ Molecular Weight: 141,1 g/mol Schmelzpunkt: 139…143 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02181193 CAS-Nummer: 402-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...