2-Fluor-6-nitrophenol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Fluoro-2-nitrophenol
PC3970-100G
PC3970-25G
APOSPC3970-100GEA
166
EUR
APOSPC3970-100G
APOSPC3970-25G
2-Fluor-6-nitrophenol 98%
2-Fluor-6-nitrophenol
Formel:
C₆H₄FNO₃ Molecular Weight: 157,1 g/mol Schmelzpunkt: 91…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042446 CAS-Nummer: 1526-17-6 EINECS: 216-199-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...