2-Aminohexafluoropropan-2-ol
Lieferant: Apollo Scientific
|
Achtung
|
PC3990-25G
PC3990-5G
APOSPC3990-25GEA
220
EUR
APOSPC3990-25G
APOSPC3990-5G
2-Aminohexafluoropropan-2-ol
2-Aminohexafluoropropan-2-ol
Formel:
C₃H₃F₆NO Schmelzpunkt: 65…67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053173 CAS-Nummer: 31253-34-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...