6-(Trifluoromethyl)pyrimidin-4-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-6-(trifluoromethyl)pyrimidine

PC400532-1G PC400532-250MG
APOSPC400532-1GEA 50.9 EUR
APOSPC400532-1G APOSPC400532-250MG
6-(Trifluoromethyl)pyrimidin-4-amine
6-(Trifluoromethyl)pyrimidin-4-amine
Formel: C5H4F3N3
Molecular Weight: 163,1000 g/mol
Schmelzpunkt: 165…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08436657
CAS-Nummer: 672-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR