Spautin-1 98+%
Lieferant: Apollo Scientific
Synonyme:
6-Fluoro-N-(4-fluorobenzyl)quinazolin-4-amine, 6-Fluoro-N-[(4-fluorophenyl)methyl]-4-quinazolinamine
PC400581-10MG
PC400581-100MG
PC400581-50MG
APOSPC400581-10MGEA
104
EUR
APOSPC400581-10MG
APOSPC400581-100MG
APOSPC400581-50MG
Spautin-1 98+%
Spautin-1
Formel:
C₁₅H₁₁F₂N₃ Molecular Weight: 271,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16962292 CAS-Nummer: 1262888-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...