6-Chloro-4-trifluoromethylpyridin-2-ylamine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-6-chloro-4-(trifluoromethyl)pyridine, 6-Chloro-4-(trifluoromethyl)pyridin-2-amine
PC400601-1G
PC400601-250MG
PC400601-5G
APOSPC400601-1GEA
222
EUR
APOSPC400601-1G
APOSPC400601-250MG
APOSPC400601-5G
6-Chloro-4-trifluoromethylpyridin-2-ylamine 95+%
6-Chloro-4-trifluoromethylpyridin-2-ylamine
Formel:
C₆H₄ClF₃N₂ Molecular Weight: 196,56 g/mol Schmelzpunkt: 45…46 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128898 CAS-Nummer: 34486-23-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...