3-Amino-6-methyl-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC400602-1G PC400602-5G
APOSPC400602-1GEA 176 EUR
APOSPC400602-1G APOSPC400602-5G
3-Amino-6-methyl-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxylic acid
3-Amino-6-methyl-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxylic acid
Formel: C₁₀H₇F₃N₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD01055887
CAS-Nummer: 610259-30-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR