(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)hydrazine
Lieferant: Apollo Scientific
|
Achtung
|
PC400621-1G
PC400621-5G
APOSPC400621-1GEA
201
EUR
APOSPC400621-1G
APOSPC400621-5G
(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)hydrazine
(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)hydrazine
Formel:
C₆H₅ClF₃N₃ Molecular Weight: 211,58 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD10568286 CAS-Nummer: 129015-68-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...