(3-Bromo-5-trifluoromethylpyridin-2-yl)dimethylamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-2-(dimethylamino)-5-(trifluoromethyl)pyridine, 3-Bromo-2-dimethylamino-5-trifluoromethylpyridine

PC400629-1G PC400629-250MG PC400629-500MG PC400629-2.5G
APOSPC400629-1GEA 243 EUR
APOSPC400629-1G APOSPC400629-250MG APOSPC400629-500MG APOSPC400629-2.5G
(3-Bromo-5-trifluoromethylpyridin-2-yl)dimethylamine
(3-Bromo-5-trifluoromethylpyridin-2-yl)dimethylamine
Formel: C₈H₈BrF₃N₂
Molecular Weight: 269,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18384845
CAS-Nummer: 216765-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR