(3-Chloro-5-trifluoromethylpyridin-2-yloxy)acetic acid ethyl ester
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2-[[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy]acetate
PC400635-1G
PC400635-250MG
PC400635-500MG
PC400635-2.5G
PC400635-5G
APOSPC400635-1GEA
366
EUR
APOSPC400635-1G
APOSPC400635-250MG
APOSPC400635-500MG
APOSPC400635-2.5G
APOSPC400635-5G
(3-Chloro-5-trifluoromethylpyridin-2-yloxy)acetic acid ethyl ester
(3-Chloro-5-trifluoromethylpyridin-2-yloxy)acetic acid ethyl ester
Formel:
C₁₀H₉ClF₃NO₃ Molecular Weight: 283.64 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18384847 CAS-Nummer: 1299607-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...