4-Fluor-2-nitrobenzaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC400738-1G
PC400738-5G
APOSPC400738-1GEA
36.4
EUR
APOSPC400738-1G
APOSPC400738-5G
4-Fluor-2-nitrobenzaldehyd 98%
4-Fluoro-2-nitrobenzaldehyde
Formel:
C₇H₄FNO₃ Molecular Weight: 169.11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01666439 CAS-Nummer: 2923-96-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...