6-Bromo-2-trifluoromethyl-1H-benzoimidazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2-(trifluoromethyl)-1H-benzimidazole

PC401035-1G PC401035-5G
APOSPC401035-1GEA 87 EUR
APOSPC401035-1G APOSPC401035-5G
6-Bromo-2-trifluoromethyl-1H-benzoimidazole
6-Bromo-2-trifluoromethyl-1H-benzoimidazole
Formel: C₈H₄BrF₃N₂
Molecular Weight: 265,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00460102
CAS-Nummer: 3671-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR