3-Fluoro-5-(trifluoromethyl)pyridine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-5-(trifluoromethyl)picolinic acid

PC401042-1G PC401042-250MG
APOSPC401042-1GEA 214 EUR
APOSPC401042-1G APOSPC401042-250MG
3-Fluoro-5-(trifluoromethyl)pyridine-2-carboxylic acid
3-Fluoro-5-(trifluoromethyl)pyridine-2-carboxylic acid
Formel: C₇H₃F₄NO₂
Molecular Weight: 209,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11036125
CAS-Nummer: 89402-28-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR