Crizotinib 95%

Lieferant: Apollo Scientific

Synonyme: (R)-3-(1-(2,6-Dichloro-3-fluorophenyl)ethoxy)-5-(1-(piperidin-4-yl)-1H-pyrazol-4-yl)pyridin-2-amine

PC402134-1G PC402134-500MG
APOSPC402134-1GEA 108 EUR
APOSPC402134-1G APOSPC402134-500MG
Crizotinib 95%
Crizotinib
Formel: C₂₁H₂₂Cl₂FN₅O
Molecular Weight: 450,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12407409
CAS-Nummer: 877399-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR