2-Bromo-5-(trifluoromethyl)-1,3,4-thiadiazole

Lieferant: Apollo Scientific
Achtung

PC4029-1G PC4029-5G PC4029-500MG
APOSPC4029-1GEA 192 EUR
APOSPC4029-1G APOSPC4029-5G APOSPC4029-500MG
2-Bromo-5-(trifluoromethyl)-1,3,4-thiadiazole
2-Bromo-5-(trifluoromethyl)-1,3,4-thiadiazole
Formel: C₃BrF₃N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00015535
CAS-Nummer: 37461-61-3
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR