(4-Fluorobenzyl)hydrazine dihydrochloride

Lieferant: Apollo Scientific

PC403065-10G PC403065-25G PC403065-5G
APOSPC403065-10GEA 539 EUR
APOSPC403065-10G APOSPC403065-25G APOSPC403065-5G
(4-Fluorobenzyl)hydrazine dihydrochloride
(4-Fluorobenzyl)hydrazine dihydrochloride
Formel: C₇H₁₁Cl₂FN₂
Molecular Weight: 213,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06800719
CAS-Nummer: 1000805-93-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR