2-Hydrazino-5-(trifluoromethyl)-1,3,4-thiadiazole

Lieferant: Apollo Scientific
Achtung

PC4034-1G PC4034-500MG
APOSPC4034-1GEA 232 EUR
APOSPC4034-1G APOSPC4034-500MG
2-Hydrazino-5-(trifluoromethyl)-1,3,4-thiadiazole
2-Hydrazino-5-(trifluoromethyl)-1,3,4-thiadiazole
Formel: C₃H₃F₃N₄S
Lagertemperatur: Kühlschrank
MDL: MFCD07786303
CAS-Nummer: 299440-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR