1-(6-Fluoropyridin-3-yl)ethanone 95+%
Lieferant: Apollo Scientific
PC404005-250MG
PC404005-1G
APOSPC404005-250MGEA
21.9
EUR
APOSPC404005-250MG
APOSPC404005-1G
1-(6-Fluoropyridin-3-yl)ethanone 95+%
1-(6-Fluoropyridin-3-yl)ethanone
Formel:
C₇H₆FNO Molecular Weight: 139,1300 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07375076 CAS-Nummer: 84331-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...