1-[Amino(4-fluorphenyl)methyl]naphthalin-2-ol Hydrochlorid

Lieferant: Apollo Scientific

PC404507-1G PC404507-250MG
APOSPC404507-1GEA 739 EUR
APOSPC404507-1G APOSPC404507-250MG
1-[Amino(4-fluorphenyl)methyl]naphthalin-2-ol Hydrochlorid
1-[Amino(4-fluorphenyl)methyl]naphthalin-2-ol Hydrochlorid
Formel: C₁₇H₁₅ClFNO
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1169968-26-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR