3,3-Dichloro-1,1,1-trifluoroacetone 97%

Lieferant: Apollo Scientific
Achtung

PC4046-25G PC4046-5G
APOSPC4046-25GEA 426 EUR
APOSPC4046-25G APOSPC4046-5G
3,3-Dichloro-1,1,1-trifluoroacetone 97%
3,3-Dichloro-1,1,1-trifluoroacetone
Formel: C₃HCl₂F₃O
Molecular Weight: 180.94 g/mol
Siedepunkt: 75…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08461592
CAS-Nummer: 126266-75-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR