2-Chlor-7-fluor-4-methylchinolin

Lieferant: Apollo Scientific

PC404619-1G PC404619-5G
APOSPC404619-1GEA 0 EUR
APOSPC404619-1G APOSPC404619-5G
2-Chlor-7-fluor-4-methylchinolin
2-Chloro-7-fluoro-4-methylquinoline
Formel: C₁₀H₇ClFN
Lagertemperatur: Raumtemperatur
CAS-Nummer: 271241-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR