2-Brom-4-(pentafluorthio)anilin 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Amino-3-bromophenylsulphur pentafluoride, (4-Amino-3-bromophenyl)pentafluorosulphur
PC405596-1G
PC405596-250MG
PC405596-5G
APOSPC405596-1GEA
345
EUR
APOSPC405596-1G
APOSPC405596-250MG
APOSPC405596-5G
2-Brom-4-(pentafluorthio)anilin 95%
2-Brom-4-(pentafluorthio)anilin
Formel:
C₆H₅BrF₅NS Molecular Weight: 298,716 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20731078 CAS-Nummer: 159727-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...