3-Brom-5-(pentafluorthio)benzoesäure

Lieferant: Apollo Scientific
Achtung

PC405651-1G PC405651-5G
APOSPC405651-1GEA 348 EUR
APOSPC405651-1G APOSPC405651-5G
3-Brom-5-(pentafluorthio)benzoesäure
3-Bromo-5-(pentafluorothio)benzoic acid
Formel: C₇H₄BrF₅O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD16652482
CAS-Nummer: 1180675-98-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR