4-(Pentafluorothio)benzenesulphonyl chloride 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC405657-1G
PC405657-5G
PC405657-10G
APOSPC405657-1GEA
342
EUR
APOSPC405657-1G
APOSPC405657-5G
APOSPC405657-10G
4-(Pentafluorothio)benzenesulphonyl chloride 97%
4-(Pentafluorothio)benzenesulphonyl chloride
Formel:
C₆H₄ClF₅O₂S₂ Molecular Weight: 302.66 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16652450 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...