3-(Pentafluorothio)benzenesulphonyl chloride 97%

Lieferant: Apollo Scientific

Synonyme: 3-(pentafluoro-lambda-sulphanyl)benzene-1-sulphonyl chloride, 3-(Pentafluoro-lambda6-sulphanyl)benzene-1-sulphonyl chloride, 3-(Pentafluorosulphur)benzenesulphonyl chloride

PC405660-1G PC405660-5G PC405660-10G
APOSPC405660-1GEA 348 EUR
APOSPC405660-1G APOSPC405660-5G APOSPC405660-10G
3-(Pentafluorothio)benzenesulphonyl chloride 97%
3-(Pentafluorothio)benzenesulphonyl chloride
Formel: C₆H₄ClF₅O₂S₂
Molecular Weight: 302,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16652436
CAS-Nummer: 1211520-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR