3-Brom-5-(pentafluorthio)benzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
PC405664-1G
PC405664-5G
APOSPC405664-1GEA
192
EUR
APOSPC405664-1G
APOSPC405664-5G
3-Brom-5-(pentafluorthio)benzaldehyd
3-Bromo-5-(pentafluorothio)benzaldehyde
Formel:
C₇H₄BrF₅OS Molecular Weight: 311,06 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16652483 CAS-Nummer: 1240257-22-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...