2-Chlor-4-(pentafluorthio)anilin

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-3-chlorophenylsulphur pentafluoride, (4-Amino-3-chlorophenyl)pentafluorosulphur

PC405667-1G PC405667-5G PC405667-10G
APOSPC405667-1GEA 1060 EUR
APOSPC405667-1G APOSPC405667-5G APOSPC405667-10G
2-Chlor-4-(pentafluorthio)anilin
2-Chlor-4-(pentafluorthio)anilin
Formel: C₆H₅ClF₅NS
Molecular Weight: 253,620 g/mol
Schmelzpunkt: 56…58 °C
Lagertemperatur: Kühlschrank
MDL: MFCD20731080
CAS-Nummer: 165114-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR