2-Fluor-4-(pentafluorthio)benzonitril 98%
Lieferant: Apollo Scientific
PC405673-5G
PC405673-1G
APOSPC405673-5GEA
1050
EUR
APOSPC405673-5G
APOSPC405673-1G
2-Fluor-4-(pentafluorthio)benzonitril 98%
2-Fluor-4-(pentafluorthio)benzonitril
Formel:
C₇H₃F₆NS Molecular Weight: 247.16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16652501 CAS-Nummer: 1240256-91-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...