3-Fluoro-5-(pentafluorothio)phenol 98%

Lieferant: Apollo Scientific

Synonyme: 3-Fluoro-5-(pentafluorosulphur)phenol

PC405683-1G PC405683-5G
APOSPC405683-1GEA 348 EUR
APOSPC405683-1G APOSPC405683-5G
3-Fluoro-5-(pentafluorothio)phenol 98%
3-Fluoro-5-(pentafluorothio)phenol
Formel: C₆H₄F₆OS
Molecular Weight: 238,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16652462
CAS-Nummer: 1240257-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR