3-Fluoro-3-oxetanemethanol

Lieferant: Apollo Scientific

Synonyme: 3-Fluoro-3-hydroxymethyloxetane, (3-Fluorooxetan-3-yl)methanol

PC406005-1G PC406005-500MG
APOSPC406005-1GEA 1050 EUR
APOSPC406005-1G APOSPC406005-500MG
3-Fluoro-3-oxetanemethanol
3-Fluoro-3-oxetanemethanol
Formel: C₄H₇FO₂
Molecular Weight: 106,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14706188
CAS-Nummer: 865451-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR