2-(2-Fluorphenyl)ethanol 99%

Lieferant: Apollo Scientific

Synonyme: 2-Fluorophenethyl alcohol

PC4066-25G
APOSPC4066-25GEA 154 EUR
APOSPC4066-25G
2-(2-Fluorphenyl)ethanol 99%
2-(2-Fluorphenyl)ethanol
Formel: C₈H₉FO
Molecular Weight: 140.16 g/mol
Dichte: 1.045 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002887
CAS-Nummer: 50919-06-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR