4-Bromo-2,3-difluorobenzeneboronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2,3-difluorophenylboronic acid

PC4069-1G PC4069-5G PC4069-10G
APOSPC4069-1GEA 29 EUR
APOSPC4069-1G APOSPC4069-5G APOSPC4069-10G
4-Bromo-2,3-difluorobenzeneboronic acid 98%
4-Bromo-2,3-difluorobenzeneboronic acid
Formel: C₆H₄BBrF₂O₂
Molecular Weight: 236,8066 g/mol
Schmelzpunkt: 225 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02093065
CAS-Nummer: 374790-99-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR