4-Brom-8-fluorchinolin

Lieferant: Apollo Scientific

PC407011-1G PC407011-5G
APOSPC407011-1GEA 82.8 EUR
APOSPC407011-1G APOSPC407011-5G
4-Brom-8-fluorchinolin
4-Bromo-8-fluoroquinoline
Formel: C₉H₅BrFN
Molecular Weight: 226.0451 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07700231
CAS-Nummer: 927800-38-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR