1-Benzyloxy-4-fluorbenzol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Benzyloxy)-4-fluorobenzene

PC4071-5G PC4071-1G
APOSPC4071-5GEA 29 EUR
APOSPC4071-5G APOSPC4071-1G
1-Benzyloxy-4-fluorbenzol 98%
1-Benzyloxy-4-fluorbenzol
Formel: C₁₃H₁₁FO
Molecular Weight: 202.23
Lagertemperatur: Raumtemperatur
MDL: MFCD03788273
CAS-Nummer: 370-78-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR