4-Bromo-2-fluorophenylboronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-fluorobenzeneboronic acid

PC4072-5G
APOSPC4072-5GEA 191 EUR
APOSPC4072-5G
4-Bromo-2-fluorophenylboronic acid 98%
4-Bromo-2-fluorophenylboronic acid
Formel: C₆H₅BBrFO₂
Molecular Weight: 218,82 g/mol
Schmelzpunkt: 238…242 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01318110
CAS-Nummer: 216393-64-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR