4-Bromo-3-fluorophenylboronic acid 95+%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-3-fluorobenzeneboronic acid

PC4073-5G PC4073-10G PC4073-1G
APOSPC4073-5GEA 231 EUR
APOSPC4073-5G APOSPC4073-10G APOSPC4073-1G
4-Bromo-3-fluorophenylboronic acid 95+%
4-Bromo-3-fluorophenylboronic acid
Formel: C₆H₅BBrFO₂
Molecular Weight: 218.82 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093066
CAS-Nummer: 374790-97-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR