2-Fluor-4-formylphenylboronsäure 95%

Lieferant: Apollo Scientific
Achtung

PC408013-25G PC408013-5G PC408013-1G
APOSPC408013-25GEA 194 EUR
APOSPC408013-25G APOSPC408013-5G APOSPC408013-1G
2-Fluor-4-formylphenylboronsäure 95%
2-Fluor-4-formylphenylboronsäure
Formel: C₇H₆BFO₃
Molecular Weight: 167,9300 g/mol
Schmelzpunkt: 161…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07367058
CAS-Nummer: 871126-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR