4-[4-Chlor-3-(trifluormethyl)phenyl]-4-piperidinol 97%

Lieferant: Apollo Scientific

Synonyme: 4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-piperidinol

PC408070-100G PC408070-25G PC408070-5G
APOSPC408070-100GEA 848 EUR
APOSPC408070-100G APOSPC408070-25G APOSPC408070-5G
4-[4-Chlor-3-(trifluormethyl)phenyl]-4-piperidinol 97%
4-[4-Chlor-3-(trifluormethyl)phenyl]-4-piperidinol
Formel: C₁₂H₁₃ClF₃NO
Molecular Weight: 279.69 g/mol
Schmelzpunkt: 138…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00066999
CAS-Nummer: 21928-50-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR