2-Chlorotetrafluoropropionic acid 95%

Lieferant: Apollo Scientific

PC408096-10G PC408096-5G
APOSPC408096-10GEA 212 EUR
APOSPC408096-10G APOSPC408096-5G
2-Chlorotetrafluoropropionic acid 95%
2-Chlorotetrafluoropropionic acid
Formel: C₃HClF₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00465559
CAS-Nummer: 6189-02-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR