6-Iodo-1,1,1,2,2-pentafluorohexane
Lieferant: Apollo Scientific
|
Achtung
|
PC408217-5G
PC408217-1G
PC408217-25G
APOSPC408217-5GEA
183
EUR
APOSPC408217-5G
APOSPC408217-1G
APOSPC408217-25G
6-Iodo-1,1,1,2,2-pentafluorohexane
6-Iodo-1,1,1,2,2-pentafluorohexane
Formel:
C₆H₈F₅I Molecular Weight: 302.03 g/mol Siedepunkt: 62…64 °C/15mmHg Dichte: 1.65 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD06247809 CAS-Nummer: 344452-10-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...