2-bromo-3-fluoro-5-methylpyridine 98%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-3-fluoro-5-methylpyridine

PC408224-100G PC408224-25G
APOSPC408224-100GEA 432 EUR
APOSPC408224-100G APOSPC408224-25G
2-bromo-3-fluoro-5-methylpyridine 98%
2-bromo-3-fluoro-5-methylpyridine
Formel: C₆H₅BrFN
Molecular Weight: 190,015 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09909840
CAS-Nummer: 34552-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR