2-Amino-5-fluor-6-methylpyridin 96%

Lieferant: Apollo Scientific

PC408225-1G PC408225-25G PC408225-5G
APOSPC408225-1GEA 75.4 EUR
APOSPC408225-1G APOSPC408225-25G APOSPC408225-5G
2-Amino-5-fluor-6-methylpyridin 96%
2-Amino-5-fluoro-6-methylpyridine
Formel: C₆H₇FN₂
Molecular Weight: 126,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09835298
CAS-Nummer: 110919-71-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR