4-Brom-3-fluorphenylessigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC408233-100G
PC408233-25G
PC408233-5G
APOSPC408233-100GEA
480
EUR
APOSPC408233-100G
APOSPC408233-25G
APOSPC408233-5G
4-Brom-3-fluorphenylessigsäure 98%
4-Brom-3-fluorphenylessigsäure
Formel:
C₈H₆BrFO₂ Molecular Weight: 233,0300 g/mol Schmelzpunkt: 111…113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12756200 CAS-Nummer: 942282-40-8 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...