4-Fluor-2-methoxybenzylalkohol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Fluoro-2-methoxyphenyl)methanol
PC408257-1G
PC408257-5G
PC408257-10G
APOSPC408257-1GEA
45.1
EUR
APOSPC408257-1G
APOSPC408257-5G
APOSPC408257-10G
4-Fluor-2-methoxybenzylalkohol 98%
4-Fluor-2-methoxybenzylalkohol
Formel:
C₈H₉FO₂ Molecular Weight: 156,16 g/mol Siedepunkt: 110…112 °C/4mmHg Schmelzpunkt: 53…55 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04116338 CAS-Nummer: 157068-03-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...