(4-Fluor-2-methoxyphenyl)essigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(4-Fluoro-2-methoxyphenyl)acetic acid
PC408263-1G
PC408263-5G
APOSPC408263-1GEA
126
EUR
APOSPC408263-1G
APOSPC408263-5G
(4-Fluor-2-methoxyphenyl)essigsäure 98%
(4-Fluor-2-methoxyphenyl)essigsäure
Formel:
C₉H₉FO₃ Schmelzpunkt: 106…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660193 CAS-Nummer: 886498-61-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...