6-fluoronicotinonitrile 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Cyano-2-fluoropyridine, 6-Fluoronicotinonitrile
PC408290-1G
PC408290-5G
APOSPC408290-1GEA
21.9
EUR
APOSPC408290-1G
APOSPC408290-5G
6-fluoronicotinonitrile 97%
6-fluoronicotinonitrile
Formel:
C₆H₃FN₂ Molecular Weight: 122,102 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09261087 CAS-Nummer: 3939-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...